Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cd37
Entrez Gene ID
548518
Gene Name
CD37 molecule
Gene Aliases
-
Chromosome Location
Chr.7: 95829923 - 95886994 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015801.2 NP_001015801.2 5 - 6 436 98
GenBank mRNA BC089757.1 - 5 - 6 436 98

View Full Product Details