Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
afg3l1p
Entrez Gene ID
548660
Gene Name
AFG3-like AAA ATPase 1, pseudogene
Gene Aliases
afg3, afg3l1
Chromosome Location
Chr.4: 61744348 - 61760587 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015906.3 NP_001015906.1 10 - 11 1300 85
GenBank mRNA BC170788.1 - 10 - 11 1299 85
BC171124.1 - 10 - 11 1299 85
CR926435.2 - 10 - 11 1300 85

View Full Product Details