Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
eif4e
Entrez Gene ID
548663
Gene Name
eukaryotic translation initiation factor 4E
Gene Aliases
cbp, eIF-4E, eif4e1, eif4el1, eif4f
Chromosome Location
Chr.1: 65531286 - 65549081 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015909.1 NP_001015909.1 4 - 5 279 101
GenBank mRNA CR926428.1 - 4 - 5 279 101

View Full Product Details