Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stc2
Entrez Gene ID
548758
Gene Name
stanniocalcin 2
Gene Aliases
-
Chromosome Location
Chr.3: 33820368 - 33832728 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016004.2 NP_001016004.1 3 - 4 649 92
GenBank mRNA BC121338.1 - 3 - 4 634 92
CR848248.2 - 3 - 4 649 92

View Full Product Details