Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fmo1
Entrez Gene ID
549120
Gene Name
flavin containing monooxygenase 1
Gene Aliases
-
Chromosome Location
Chr.4: 97558397 - 97577515 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016366.2 NP_001016366.1 6 - 7 776 78
XM_012961090.2 XP_012816544.1 6 - 7 820 78
GenBank mRNA BC153695.1 - 5 - 6 686 78
CR761286.2 - 6 - 7 776 78

View Full Product Details