Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oat
Entrez Gene ID
549690
Gene Name
ornithine aminotransferase
Gene Aliases
hoga, oat.1
Chromosome Location
Chr.7: 11393760 - 11406503 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016936.2 NP_001016936.1 2 - 3 248 62
GenBank mRNA BC167123.1 - 1 - 2 211 62
BC167345.1 - 1 - 2 487 62
CR762076.2 - 2 - 3 248 62

View Full Product Details