Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
poldip2
Entrez Gene ID
549852
Gene Name
polymerase (DNA-directed), delta interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.2: 27377117 - 27386708 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017098.1 NP_001017098.1 2 - 3 259 66
XM_012957030.2 XP_012812484.1 2 - 3 256 66
XM_018091275.1 XP_017946764.1 2 - 3 262 66
GenBank mRNA BC118740.1 - 2 - 3 243 66
BC135627.1 - 2 - 3 239 66
BC167512.1 - 2 - 3 243 66
CR761305.1 - 2 - 3 259 66

View Full Product Details