Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ldlrap1
Entrez Gene ID
549868
Gene Name
low density lipoprotein receptor adaptor protein 1
Gene Aliases
arh, arh1, arh2, fhcb1, fhcb2, ldlrap1-a, ldlrap1-b, xptb
Chromosome Location
Chr.2: 85410914 - 85426247 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017114.2 NP_001017114.1 8 - 9 845 63
GenBank mRNA BC157335.1 - 8 - 9 857 63
BC158170.1 - 8 - 9 857 63
CR761192.2 - 8 - 9 845 63

View Full Product Details