Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ifi30
Entrez Gene ID
549950
Gene Name
interferon, gamma-inducible protein 30
Gene Aliases
GILT
Chromosome Location
Chr.1: 84907018 - 84926541 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017196.2 NP_001017196.1 4 - 5 487 107
XM_012963420.1 XP_012818874.1 4 - 5 657 107
GenBank mRNA CR760545.2 - 4 - 5 487 107

View Full Product Details