Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ifi30
Entrez Gene ID
549950
Gene Name
interferon, gamma-inducible protein 30
Gene Aliases
GILT
Chromosome Location
Chr.1: 84907018 - 84926541 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017196.2 NP_001017196.1 6 - 7 710 90
XM_012963420.1 XP_012818874.1 6 - 7 880 90
GenBank mRNA CR760545.2 - 6 - 7 710 90

View Full Product Details