Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hist1h2bj
Entrez Gene ID
550005
Gene Name
histone cluster 1, H2bj
Gene Aliases
h2bfr
Chromosome Location
Chr.Un|NW_016683463: 48117 - 48750 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017251.2 NP_001017251.1 1 - 1 453 65
GenBank mRNA CR760086.2 - 1 - 1 453 65

View Full Product Details