Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsbn1l
Entrez Gene ID
550058
Gene Name
round spermatid basic protein 1 like
Gene Aliases
-
Chromosome Location
Chr.3: 68717348 - 68743325 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017304.2 NP_001017304.1 6 - 7 1691 109
GenBank mRNA CR855556.2 - 6 - 7 1691 109

View Full Product Details