Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atp23
Entrez Gene ID
594990
Gene Name
ATP23 metallopeptidase and ATP synthase assembly factor homolog
Gene Aliases
xrcc6bp1
Chromosome Location
Chr.3: 53799923 - 53807336 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030431.1 NP_001025602.1 1 - 2 204 80
XM_012959130.2 XP_012814584.1 1 - 2 444 80
GenBank mRNA BC091052.1 - 1 - 2 204 80
CR761179.2 - 1 - 2 179 80

View Full Product Details