The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011348.2 | NP_001011348.2 | 2 - 3 | 1043 | 94 |
GenBank mRNA | BC088505.1 | - | 2 - 3 | 748 | 94 |
CR848474.2 | - | 2 - 3 | 1043 | 94 |