Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tmem242
Entrez Gene ID
548795
Gene Name
transmembrane protein 242
Gene Aliases
c6orf35
Chromosome Location
Chr.5: 41683523 - 41706430 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016041.2 NP_001016041.1 1 - 2 109 69
GenBank mRNA BC170837.1 - 1 - 2 109 69
BC170839.1 - 1 - 2 109 69
CR848094.2 - 1 - 2 109 69

View Full Product Details