Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rad1
Entrez Gene ID
548799
Gene Name
RAD1 checkpoint DNA exonuclease
Gene Aliases
xrad1
Chromosome Location
Chr.1: 185965149 - 185970594 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016045.2 NP_001016045.1 4 - 5 942 77
XM_012961937.2 XP_012817391.1 5 - 6 762 77
XM_012961942.2 XP_012817396.1 4 - 5 951 77
GenBank mRNA BC157217.1 - 4 - 5 869 77
CR762361.2 - 4 - 5 942 77

View Full Product Details