Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rad1
Entrez Gene ID
548799
Gene Name
RAD1 checkpoint DNA exonuclease
Gene Aliases
xrad1
Chromosome Location
Chr.1: 185965149 - 185970594 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016045.2 NP_001016045.1 5 - 6 1029 75
XM_012961937.2 XP_012817391.1 6 - 7 849 75
XM_012961942.2 XP_012817396.1 5 - 6 1038 75
GenBank mRNA BC157217.1 - 5 - 6 956 75
CR762361.2 - 5 - 6 1029 75

View Full Product Details