Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nus1
Entrez Gene ID
548844
Gene Name
NUS1, dehydrodolichyl diphosphate synthase subunit
Gene Aliases
ngbr
Chromosome Location
Chr.5: 70037580 - 70046985 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016090.2 NP_001016090.1 2 - 3 567 66
XM_018093874.1 XP_017949363.1 2 - 3 761 66
GenBank mRNA BC121526.1 - 2 - 3 566 66
CR762183.2 - 2 - 3 567 66

View Full Product Details