Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbxl3
Entrez Gene ID
548882
Gene Name
F-box and leucine-rich repeat protein 3
Gene Aliases
-
Chromosome Location
Chr.2: 122882934 - 122900819 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016128.2 NP_001016128.1 3 - 4 761 65
GenBank mRNA BC166143.1 - 3 - 4 762 65
BC170590.1 - 3 - 4 577 65
CR762059.2 - 3 - 4 761 65

View Full Product Details