Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cox18
Entrez Gene ID
549087
Gene Name
cytochrome c oxidase assembly factor COX18
Gene Aliases
-
Chromosome Location
Chr.Un|NW_016683393: 578738 - 587720 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016333.2 NP_001016333.1 4 - 5 1139 99
XM_012962186.2 XP_012817640.1 5 - 6 981 99
XM_012962189.2 XP_012817643.1 5 - 6 980 99
XM_012962191.1 XP_012817645.1 5 - 6 897 99
XM_018089476.1 XP_017944965.1 4 - 5 647 99
GenBank mRNA BC171112.1 - 4 - 5 1051 99
CR761413.2 - 4 - 5 1139 99

View Full Product Details