Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tmem18
Entrez Gene ID
549089
Gene Name
transmembrane protein 18
Gene Aliases
-
Chromosome Location
Chr.5: 129801043 - 129805446 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016335.2 NP_001016335.1 3 - 4 270 107
GenBank mRNA BC170719.1 - 3 - 4 258 107
BC170723.1 - 3 - 4 258 107
CR761408.2 - 3 - 4 270 107

View Full Product Details