Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pgk1
Entrez Gene ID
549299
Gene Name
phosphoglycerate kinase 1
Gene Aliases
pgk2, pgka
Chromosome Location
Chr.8: 38671742 - 38686207 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016545.2 NP_001016545.1 6 - 7 654 113
XM_012968421.1 XP_012823875.1 6 - 7 611 113
XM_012968422.1 XP_012823876.1 6 - 7 742 113
GenBank mRNA CR760747.2 - 6 - 7 654 113

View Full Product Details