Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cplane2
Entrez Gene ID
549439
Gene Name
ciliogenesis and planar polarity effector 2
Gene Aliases
c1orf89, rsg1
Chromosome Location
Chr.Un|NW_016688196: 7 - 3297 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016685.2 NP_001016685.1 2 - 3 368 100
GenBank mRNA BC170963.1 - 2 - 3 317 100
CR760197.2 - 2 - 3 368 100

View Full Product Details