Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btf3
Entrez Gene ID
549451
Gene Name
basic transcription factor 3
Gene Aliases
beta-nac, btf3a, btf3b, nacb
Chromosome Location
Chr.1: 166487922 - 166493753 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016697.2 NP_001016697.1 5 - 6 598 94
XM_018093422.1 XP_017948911.1 5 - 6 600 94
GenBank mRNA CR760165.2 - 5 - 6 598 94

View Full Product Details