Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
zbtb8os
Entrez Gene ID
549545
Gene Name
zinc finger and BTB domain containing 8 opposite strand
Gene Aliases
arch, arch2
Chromosome Location
Chr.2: 86190955 - 86199276 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016791.2 NP_001016791.1 4 - 5 379 105
XM_012956998.2 XP_012812452.1 4 - 5 348 105
GenBank mRNA CR855646.2 - 4 - 5 379 105

View Full Product Details