Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cmss1
Entrez Gene ID
549697
Gene Name
cms1 ribosomal small subunit homolog
Gene Aliases
c3orf26
Chromosome Location
Chr.2: 30542427 - 30737171 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016943.2 NP_001016943.1 5 - 6 445 84
GenBank mRNA BC135225.1 - 5 - 6 499 84
CR761873.2 - 5 - 6 445 84

View Full Product Details