Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prss8
Entrez Gene ID
549734
Gene Name
serine protease 8
Gene Aliases
cap1, prostasin
Chromosome Location
Chr.9: 1857757 - 1868019 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016980.2 NP_001016980.1 4 - 5 511 89
GenBank mRNA CR848351.2 - 4 - 5 511 89

View Full Product Details