Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mpped2
Entrez Gene ID
549855
Gene Name
metallophosphoesterase domain containing 2
Gene Aliases
239fb
Chromosome Location
Chr.4: 10368657 - 10464298 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017101.2 NP_001017101.1 6 - 7 1020 66
XM_012961172.2 XP_012816626.1 6 - 7 1162 66
GenBank mRNA CR761291.2 - 6 - 7 1020 66

View Full Product Details