Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zfand5
Entrez Gene ID
549882
Gene Name
zinc finger AN1-type containing 5
Gene Aliases
za20d2, zfand5-a, zfand5-b, zfand5a, znf216
Chromosome Location
Chr.1: 118274399 - 118291176 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017128.2 NP_001017128.1 4 - 5 399 97
XM_012963336.2 XP_012818790.1 4 - 5 785 97
XM_012963340.2 XP_012818794.1 4 - 5 679 97
GenBank mRNA BC123970.1 - 4 - 5 393 97
CR761102.2 - 4 - 5 399 97

View Full Product Details