Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
zfand5
Entrez Gene ID
549882
Gene Name
zinc finger AN1-type containing 5
Gene Aliases
za20d2, zfand5-a, zfand5-b, zfand5a, znf216
Chromosome Location
Chr.1: 118274399 - 118291176 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017128.2 NP_001017128.1 5 - 6 522 104
XM_012963336.2 XP_012818790.1 5 - 6 908 104
XM_012963340.2 XP_012818794.1 5 - 6 802 104
GenBank mRNA BC123970.1 - 5 - 6 516 104
BC166153.1 - 5 - 6 757 104
CR761102.2 - 5 - 6 522 104

View Full Product Details