Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
endog
Entrez Gene ID
549956
Gene Name
endonuclease G
Gene Aliases
-
Chromosome Location
Chr.8: 26343513 - 26347347 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017202.2 NP_001017202.1 1 - 2 504 82
GenBank mRNA CR760515.2 - 1 - 2 504 82

View Full Product Details