Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
qpct
Entrez Gene ID
549999
Gene Name
glutaminyl-peptide cyclotransferase
Gene Aliases
qpctl
Chromosome Location
Chr.5: 59086858 - 59109960 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017245.2 NP_001017245.1 1 - 2 134 109
GenBank mRNA BC118705.1 - 1 - 2 178 109
CR760147.2 - 1 - 2 134 109

View Full Product Details