Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
qpct
Entrez Gene ID
549999
Gene Name
glutaminyl-peptide cyclotransferase
Gene Aliases
qpctl
Chromosome Location
Chr.5: 59086858 - 59109960 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017245.2 NP_001017245.1 3 - 4 559 88
GenBank mRNA BC118705.1 - 3 - 4 603 88
CR760147.2 - 3 - 4 559 88

Gene Symbol
LOC108644434
Entrez Gene ID
108644434
Gene Name
glutaminyl-peptide cyclotransferase-like
Gene Aliases
-
Chromosome Location
Chr.5: 59120273 - 59140491 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018094262.1 XP_017949751.1 3 - 4 553 88

View Full Product Details