Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
pard6g
Entrez Gene ID
550092
Gene Name
par-6 family cell polarity regulator gamma
Gene Aliases
par-6g, par6gamma, pard6b
Chromosome Location
Chr.6: 29494060 - 29558772 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017338.2 NP_001017338.1 2 - 3 391 85
GenBank mRNA CR855473.2 - 2 - 3 391 85

View Full Product Details