Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tuba3c
Entrez Gene ID
548688
Gene Name
tubulin, alpha 3c
Gene Aliases
tuba2, tuba3d
Chromosome Location
Chr.2: 49242150 - 49248847 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015934.4 NP_001015934.1 - 1107 90
GenBank mRNA BC080160.1 - 2 - 3 357 90
BC135250.1 - 2 - 3 339 90
CR926373.2 - 2 - 3 327 90

View Full Product Details