Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lysmd2
Entrez Gene ID
733450
Gene Name
LysM, putative peptidoglycan-binding, domain containing 2
Gene Aliases
-
Chromosome Location
Chr.3: 92437436 - 92447347 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044403.2 NP_001037868.1 2 - 3 817 66
GenBank mRNA BC107589.1 - 2 - 3 637 66
BC118885.1 - 2 - 3 610 66
CU075816.1 - 2 - 3 606 66

View Full Product Details