Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
bola3
Entrez Gene ID
733564
Gene Name
bolA family member 3
Gene Aliases
-
Chromosome Location
Chr.3: 112954200 - 112960640 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044475.1 NP_001037940.1 1 - 2 59 73
GenBank mRNA BC135597.1 - 1 - 2 101 73
CR761390.2 - 1 - 2 59 73

View Full Product Details