Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hint3
Entrez Gene ID
733971
Gene Name
histidine triad nucleotide binding protein 3
Gene Aliases
-
Chromosome Location
Chr.5: 47230685 - 47237606 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045681.1 NP_001039146.1 2 - 3 291 72
GenBank mRNA BC158181.1 - 2 - 3 342 72
CR942552.2 - 2 - 3 291 72

View Full Product Details