Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
eln1
Entrez Gene ID
779443
Gene Name
tropoelastin 1
Gene Aliases
eln
Chromosome Location
Chr.2: 37685140 - 37763602 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078709.1 NP_001072177.1 15 - 16 915 73
XM_012957183.2 XP_012812637.1 17 - 18 1101 73
XM_012957184.2 XP_012812638.1 16 - 17 1071 73
XM_012957185.2 XP_012812639.1 17 - 18 1101 73
XM_012957188.1 XP_012812642.1 17 - 18 1030 73
XM_012957189.2 XP_012812643.1 15 - 16 1008 73
XM_012957190.2 XP_012812644.1 15 - 16 980 73
XM_012957191.2 XP_012812645.1 17 - 18 1102 73
XM_012957192.2 XP_012812646.1 17 - 18 1102 73
XM_012957193.1 XP_012812647.1 17 - 18 1103 73
XM_012957194.2 XP_012812648.1 17 - 18 1101 73
XM_012957195.2 XP_012812649.1 14 - 15 950 73
XM_012957196.1 XP_012812650.1 17 - 18 1102 73
XM_012957197.1 XP_012812651.1 17 - 18 1102 73
XM_012957199.2 XP_012812653.1 13 - 14 887 73
XM_012957200.1 XP_012812654.1 17 - 18 1103 73
XM_012957201.2 XP_012812655.1 17 - 18 1102 73
XM_012957202.1 XP_012812656.1 17 - 18 1102 73
XM_012957204.1 XP_012812658.1 17 - 18 1103 73
XM_012957205.1 XP_012812659.1 17 - 18 1102 73
XM_012957206.1 XP_012812660.1 17 - 18 1102 73
XM_012957207.1 XP_012812661.1 17 - 18 1103 73
XM_012957208.1 XP_012812662.1 17 - 18 1103 73
XM_012957209.1 XP_012812663.1 17 - 18 1103 73
XM_012957210.2 XP_012812664.1 17 - 18 1103 73
XM_012957211.1 XP_012812665.1 17 - 18 1103 73
XM_018091327.1 XP_017946816.1 17 - 18 1103 73
GenBank mRNA DQ523561.1 - 15 - 16 915 73

View Full Product Details