Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
eln1
Entrez Gene ID
779443
Gene Name
tropoelastin 1
Gene Aliases
eln
Chromosome Location
Chr.2: 37685140 - 37763602 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078709.1 NP_001072177.1 44 - 45 2976 86
XM_012957183.2 XP_012812637.1 47 - 48 3195 86
XM_012957184.2 XP_012812638.1 46 - 47 3165 86
XM_012957185.2 XP_012812639.1 46 - 47 3162 86
XM_012957188.1 XP_012812642.1 47 - 48 3124 86
XM_012957189.2 XP_012812643.1 45 - 46 3102 86
XM_012957190.2 XP_012812644.1 45 - 46 3074 86
XM_012957191.2 XP_012812645.1 45 - 46 3067 86
XM_012957193.1 XP_012812647.1 47 - 48 3197 86
XM_012957194.2 XP_012812648.1 45 - 46 3048 86
XM_012957195.2 XP_012812649.1 44 - 45 3044 86
XM_012957196.1 XP_012812650.1 45 - 46 3034 86
XM_012957197.1 XP_012812651.1 45 - 46 3004 86
XM_012957199.2 XP_012812653.1 43 - 44 2981 86
XM_012957201.2 XP_012812655.1 43 - 44 2908 86
XM_012957202.1 XP_012812656.1 43 - 44 2896 86
XM_012957205.1 XP_012812659.1 43 - 44 2890 86
XM_012957206.1 XP_012812660.1 43 - 44 2875 86
XM_012957209.1 XP_012812663.1 39 - 40 2630 86
XM_018091327.1 XP_017946816.1 41 - 42 2762 86
GenBank mRNA DQ523561.1 - 44 - 45 2976 86

View Full Product Details