Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
frmd7
Entrez Gene ID
779686
Gene Name
FERM domain containing 7
Gene Aliases
-
Chromosome Location
Chr.8: 46277217 - 46292719 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018096974.1 XP_017952463.1 5 - 6 640 62
GenBank mRNA BC118833.1 - 5 - 6 394 62

View Full Product Details