Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ntn1
Entrez Gene ID
779839
Gene Name
netrin 1
Gene Aliases
Netrin-1, ntn1l, unc-6
Chromosome Location
Chr.7: 97552309 - 97622345 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078917.1 NP_001072385.1 5 - 6 1578 60
GenBank mRNA BC121568.1 - 5 - 6 1578 60

View Full Product Details