Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ppp2r3b
Entrez Gene ID
780131
Gene Name
protein phosphatase 2 regulatory subunit B', beta
Gene Aliases
-
Chromosome Location
Chr.2: 111547939 - 111588347 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079206.1 NP_001072674.1 11 - 12 1406 78
XM_012957298.2 XP_012812752.1 10 - 11 1633 78
GenBank mRNA BC124001.1 - 11 - 12 1406 78

View Full Product Details