Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
shroom4
Entrez Gene ID
780201
Gene Name
shroom family member 4
Gene Aliases
shap, shrm4
Chromosome Location
Chr.8: 15991573 - 16059640 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079276.1 NP_001072744.1 1 - 2 203 83
XM_018096333.1 XP_017951822.1 1 - 2 275 83
GenBank mRNA BC125736.1 - 1 - 2 203 83

View Full Product Details