Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
shroom4
Entrez Gene ID
780201
Gene Name
shroom family member 4
Gene Aliases
shap, shrm4
Chromosome Location
Chr.8: 15991573 - 16059640 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079276.1 NP_001072744.1 6 - 7 4192 88
XM_018096333.1 XP_017951822.1 7 - 8 4345 88
XM_018096334.1 XP_017951823.1 8 - 9 4411 88
GenBank mRNA BC125736.1 - 6 - 7 4192 88

View Full Product Details