Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cdk15
Entrez Gene ID
780381
Gene Name
cyclin-dependent kinase 15
Gene Aliases
als2cr7, pftk2
Chromosome Location
Chr.9: 47080656 - 47180732 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079451.1 NP_001072919.1 14 - 15 1264 95
XM_012970593.1 XP_012826047.1 14 - 15 1498 95
GenBank mRNA BC121427.1 - 14 - 15 1264 95

View Full Product Details