Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
derl2
Entrez Gene ID
548851
Gene Name
derlin 2
Gene Aliases
-
Chromosome Location
Chr.2: 98079229 - 98088833 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016097.3 NP_001016097.1 6 - 7 623 103
GenBank mRNA BC135880.1 - 6 - 7 623 103
CR762155.2 - 6 - 7 642 103

View Full Product Details