Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
tomm22
Entrez Gene ID
734118
Gene Name
translocase of outer mitochondrial membrane 22 homolog
Gene Aliases
tom22
Chromosome Location
Chr.4: 110048043 - 110057841 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045787.1 NP_001039252.1 2 - 3 349 92
GenBank mRNA CR942798.2 - 2 - 3 349 92

View Full Product Details