Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dhrs12
Entrez Gene ID
780002
Gene Name
dehydrogenase/reductase (SDR family) member 12
Gene Aliases
-
Chromosome Location
Chr.2: 147108382 - 147117154 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079079.1 NP_001072547.1 6 - 7 517 97
GenBank mRNA BC122052.1 - 6 - 7 517 97

View Full Product Details