Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ostm1
Entrez Gene ID
780037
Gene Name
osteopetrosis associated transmembrane protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 75626005 - 75639132 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079114.1 NP_001072582.1 3 - 4 849 84
GenBank mRNA BC122941.1 - 3 - 4 849 84
CT485681.1 - 3 - 4 835 84

View Full Product Details